Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609770 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAML PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JAML - junction adhesion molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098526.1 | 1078 | Missense Mutation | ATA,ATG | I,M 322 | NP_001091996.1 | |
NM_001286570.1 | 1078 | Missense Mutation | ATA,ATG | I,M 283 | NP_001273499.1 | |
NM_001286571.1 | 1078 | Missense Mutation | ATA,ATG | I,M 283 | NP_001273500.1 | |
NM_153206.2 | 1078 | Missense Mutation | ATA,ATG | I,M 312 | NP_694938.2 | |
XM_005271407.3 | 1078 | Missense Mutation | ATA,ATG | I,M 312 | XP_005271464.1 | |
XM_017017213.1 | 1078 | Missense Mutation | ATA,ATG | I,M 344 | XP_016872702.1 | |
XM_017017214.1 | 1078 | Missense Mutation | ATA,ATG | I,M 189 | XP_016872703.1 | |
XM_017017215.1 | 1078 | Missense Mutation | ATA,ATG | I,M 189 | XP_016872704.1 |
Set Membership: |
HapMap |