Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611665 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP73 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFAP73 - cilia and flagella associated protein 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144872.1 | 1169 | UTR 3 | NP_001138344.1 | |||
XM_011538327.2 | 1169 | Intron | XP_011536629.1 |
DDX54 - DEAD-box helicase 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111322.1 | 1169 | Silent Mutation | GGC,GGT | G,G 865 | NP_001104792.1 | |
NM_024072.3 | 1169 | Silent Mutation | GGC,GGT | G,G 864 | NP_076977.3 |
MIR7106 - microRNA 7106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |