Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 238330 MIM: 607894 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCSH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GCSH - glycine cleavage system protein H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD1L2 - polycystin 1 like 2 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076780.1 | Intron | NP_001070248.1 | ||||
NM_001278423.1 | Intron | NP_001265352.1 | ||||
NM_001278425.1 | Intron | NP_001265354.1 | ||||
NM_052892.3 | Intron | NP_443124.3 |
Set Membership: |
HapMap |