Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612234 MIM: 612235 MIM: 612333 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALHM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CALHM1 - calcium homeostasis modulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CALHM2 - calcium homeostasis modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015916.4 | 1316 | Silent Mutation | NP_057000.2 | |||
XM_006717880.2 | 1316 | Silent Mutation | XP_006717943.1 | |||
XM_006717883.2 | 1316 | Silent Mutation | XP_006717946.1 | |||
XM_006717884.3 | 1316 | UTR 3 | XP_006717947.1 | |||
XM_011539848.2 | 1316 | Silent Mutation | XP_011538150.1 | |||
XM_017016306.1 | 1316 | Silent Mutation | XP_016871795.1 | |||
XM_017016307.1 | 1316 | Silent Mutation | XP_016871796.1 | |||
XM_017016308.1 | 1316 | UTR 3 | XP_016871797.1 | |||
XM_017016309.1 | 1316 | UTR 3 | XP_016871798.1 |
PDCD11 - programmed cell death 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |