Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607185 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SEC24C - SEC24 homolog C, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004922.3 | 473 | Silent Mutation | TAC,TAT | Y,Y 28 | NP_004913.2 | |
NM_198597.2 | 473 | Silent Mutation | TAC,TAT | Y,Y 28 | NP_940999.1 | |
XM_011540380.2 | 473 | Silent Mutation | TAC,TAT | Y,Y 28 | XP_011538682.1 | |
XM_011540381.2 | 473 | Silent Mutation | TAC,TAT | Y,Y 28 | XP_011538683.1 | |
XM_017016967.1 | 473 | Missense Mutation | ACG,ATG | T,M 1 | XP_016872456.1 | |
XM_017016968.1 | 473 | Missense Mutation | ACG,ATG | T,M 1 | XP_016872457.1 |
Set Membership: |
HapMap Validated |