Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTGTTATTATTTCATTTTGAT[C/T]ATTTTAAAATATATTCTTGGTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616616 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSGALNACT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSGALNACT2 - chondroitin sulfate N-acetylgalactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319654.1 | Intron | NP_001306583.1 | ||||
NM_001319656.1 | Intron | NP_001306585.1 | ||||
NM_018590.4 | Intron | NP_061060.3 | ||||
XM_011539938.1 | Intron | XP_011538240.1 | ||||
XM_017016388.1 | Intron | XP_016871877.1 | ||||
XM_017016389.1 | Intron | XP_016871878.1 | ||||
XM_017016390.1 | Intron | XP_016871879.1 | ||||
XM_017016391.1 | Intron | XP_016871880.1 |
Set Membership: |
HapMap |