Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611360 MIM: 174763 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCI PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FANCI - Fanconi anemia complementation group I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113378.1 | Intron | NP_001106849.1 | ||||
NM_018193.2 | Intron | NP_060663.2 | ||||
XM_011521756.2 | Intron | XP_011520058.1 | ||||
XM_011521757.2 | Intron | XP_011520059.1 | ||||
XM_011521764.2 | Intron | XP_011520066.1 | ||||
XM_011521767.2 | Intron | XP_011520069.1 |
MIR6766 - microRNA 6766 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLG - DNA polymerase gamma, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126131.1 | Intron | NP_001119603.1 | ||||
NM_002693.2 | Intron | NP_002684.1 |
Set Membership: |
HapMap |