Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192240 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VEGFA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VEGFA - vascular endothelial growth factor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025366.2 | Intron | NP_001020537.2 | ||||
NM_001025367.2 | Intron | NP_001020538.2 | ||||
NM_001025368.2 | Intron | NP_001020539.2 | ||||
NM_001025369.2 | Intron | NP_001020540.2 | ||||
NM_001025370.2 | Intron | NP_001020541.2 | ||||
NM_001033756.2 | Intron | NP_001028928.1 | ||||
NM_001171622.1 | Intron | NP_001165093.1 | ||||
NM_001171623.1 | Intron | NP_001165094.1 | ||||
NM_001171624.1 | Intron | NP_001165095.1 | ||||
NM_001171625.1 | Intron | NP_001165096.1 | ||||
NM_001171626.1 | Intron | NP_001165097.1 | ||||
NM_001171627.1 | Intron | NP_001165098.1 | ||||
NM_001171628.1 | Intron | NP_001165099.1 | ||||
NM_001171629.1 | Intron | NP_001165100.1 | ||||
NM_001171630.1 | Intron | NP_001165101.1 | ||||
NM_001204384.1 | Intron | NP_001191313.1 | ||||
NM_001204385.1 | Intron | NP_001191314.1 | ||||
NM_001287044.1 | Intron | NP_001273973.1 | ||||
NM_001317010.1 | Intron | NP_001303939.1 | ||||
NM_003376.5 | Intron | NP_003367.4 |
Set Membership: |
HapMap |