Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTTGGGCAAGAAGGGAGAAAAG[C/T]GACCCAGCAGGAAGAACTTCCAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162640 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986777 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107986777 - uncharacterized LOC107986777 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017012910.1 | Intron | XP_016868399.1 | ||||
XM_017012911.1 | Intron | XP_016868400.1 |
NPY - neuropeptide Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000905.3 | Intron | NP_000896.1 |
Set Membership: |
HapMap |