Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609831 MIM: 176763 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMACHC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMACHC - methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDX1 - peroxiredoxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202431.1 | Intron | NP_001189360.1 | ||||
NM_002574.3 | Intron | NP_002565.1 | ||||
NM_181696.2 | Intron | NP_859047.1 | ||||
NM_181697.2 | Intron | NP_859048.1 |
Set Membership: |
HapMap |