Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612515 MIM: 609525 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCAF17 - DDB1 and CUL4 associated factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164821.1 | Intron | NP_001158293.1 | ||||
NM_025000.3 | Intron | NP_079276.2 | ||||
XM_006712767.1 | Intron | XP_006712830.1 | ||||
XM_006712768.1 | Intron | XP_006712831.1 | ||||
XM_011511881.1 | Intron | XP_011510183.1 | ||||
XM_011511882.1 | Intron | XP_011510184.1 | ||||
XM_017004995.1 | Intron | XP_016860484.1 | ||||
XM_017004996.1 | Intron | XP_016860485.1 | ||||
XM_017004997.1 | Intron | XP_016860486.1 | ||||
XM_017004998.1 | Intron | XP_016860487.1 | ||||
XM_017004999.1 | Intron | XP_016860488.1 | ||||
XM_017005000.1 | Intron | XP_016860489.1 | ||||
XM_017005001.1 | Intron | XP_016860490.1 | ||||
XM_017005002.1 | Intron | XP_016860491.1 |
METTL8 - methyltransferase like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |