Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614412 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCAIM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCAIM - T-cell activation inhibitor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029839.2 | Intron | NP_001025010.1 | ||||
NM_001029840.2 | Intron | NP_001025011.1 | ||||
NM_001282913.1 | Intron | NP_001269842.1 | ||||
NM_001282914.1 | Intron | NP_001269843.1 | ||||
NM_001282915.1 | Intron | NP_001269844.1 | ||||
NM_173826.3 | Intron | NP_776187.2 | ||||
XM_017006251.1 | Intron | XP_016861740.1 |
TOPAZ1 - testis and ovary specific PAZ domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |