Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116899 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDKN1A - cyclin dependent kinase inhibitor 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000389.4 | 53 | Intron | NP_000380.1 | |||
NM_001220777.1 | 53 | Intron | NP_001207706.1 | |||
NM_001220778.1 | 53 | Intron | NP_001207707.1 | |||
NM_001291549.1 | 53 | Missense Mutation | GAT,GGT | D,G 28 | NP_001278478.1 | |
NM_078467.2 | 53 | UTR 5 | NP_510867.1 |
PANDAR - promoter of CDKN1A antisense DNA damage activated RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |