Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603136 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CUL3 - cullin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257197.1 | 6367 | UTR 3 | NP_001244126.1 | |||
NM_001257198.1 | 6367 | UTR 3 | NP_001244127.1 | |||
NM_003590.4 | 6367 | UTR 3 | NP_003581.1 | |||
XM_006712800.2 | 6367 | Intron | XP_006712863.2 | |||
XM_011511994.2 | 6367 | Intron | XP_011510296.1 | |||
XM_011511995.1 | 6367 | Intron | XP_011510297.1 | |||
XM_011511996.2 | 6367 | Intron | XP_011510298.1 |
Set Membership: |
HapMap |