Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609175 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DSN1 - DSN1 homolog, MIS12 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145315.1 | Intron | NP_001138787.1 | ||||
NM_001145316.1 | Intron | NP_001138788.1 | ||||
NM_001145317.1 | Intron | NP_001138789.1 | ||||
NM_001145318.1 | Intron | NP_001138790.1 | ||||
NM_024918.3 | Intron | NP_079194.3 | ||||
XM_006723876.2 | Intron | XP_006723939.1 | ||||
XM_017028070.1 | Intron | XP_016883559.1 | ||||
XM_017028071.1 | Intron | XP_016883560.1 | ||||
XM_017028072.1 | Intron | XP_016883561.1 |