Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607471 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCAS4 - breast carcinoma amplified sequence 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010974.2 | Intron | NP_001010974.1 | ||||
NM_017843.4 | Intron | NP_060313.3 | ||||
NM_198799.3 | Intron | NP_942094.2 | ||||
XM_011528886.2 | Intron | XP_011527188.1 | ||||
XM_011528887.2 | Intron | XP_011527189.1 | ||||
XM_017027932.1 | Intron | XP_016883421.1 |
Set Membership: |
HapMap |