Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACAAGATCTACCTCTCCACCTC[C/T]TCCTCCAAGTTCTATGACCTGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607352 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984124 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107984124 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160036.1 | 5728 | Silent Mutation | TCC,TCT | S,S 310 | NP_001153508.1 | |
NM_001160037.1 | 5728 | Silent Mutation | TCC,TCT | S,S 295 | NP_001153509.1 | |
NM_015178.2 | 5728 | Silent Mutation | TCC,TCT | S,S 288 | NP_055993.2 | |
XM_017013251.1 | 5728 | Silent Mutation | TCC,TCT | S,S 310 | XP_016868740.1 |
Set Membership: |
HapMap |