Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611282 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNMBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNMBP - dynamin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318326.1 | 4736 | UTR 3 | NP_001305255.1 | |||
NM_001318327.1 | 4736 | UTR 3 | NP_001305256.1 | |||
NM_015221.3 | 4736 | UTR 3 | NP_056036.1 | |||
XM_006717735.2 | 4736 | UTR 3 | XP_006717798.1 | |||
XM_011539559.2 | 4736 | UTR 3 | XP_011537861.1 |