Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602269 MIM: 616830 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARVCF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARVCF - armadillo repeat gene deleted in velocardiofacial syndrome | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001670.2 | 182 | Intron | NP_001661.1 | |||
XM_005261242.2 | 182 | Intron | XP_005261299.1 | |||
XM_005261243.3 | 182 | Intron | XP_005261300.1 | |||
XM_005261244.3 | 182 | Intron | XP_005261301.1 | |||
XM_006724243.2 | 182 | Intron | XP_006724306.1 | |||
XM_006724245.2 | 182 | Intron | XP_006724308.1 | |||
XM_006724246.3 | 182 | Intron | XP_006724309.1 | |||
XM_006724247.3 | 182 | Intron | XP_006724310.1 | |||
XM_006724248.3 | 182 | Intron | XP_006724311.1 | |||
XM_006724249.3 | 182 | Intron | XP_006724312.1 | |||
XM_006724250.3 | 182 | Intron | XP_006724313.1 | |||
XM_011530179.2 | 182 | Intron | XP_011528481.1 | |||
XM_011530180.1 | 182 | Intron | XP_011528482.1 | |||
XM_011530181.1 | 182 | Intron | XP_011528483.1 | |||
XM_011530182.2 | 182 | Intron | XP_011528484.1 | |||
XM_011530183.2 | 182 | Intron | XP_011528485.1 |
Set Membership: |
HapMap |