Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGACCGTCTCTCCAGTCGGAC[A/G]TGCAGCTCCTTCAGGAGCCCTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611986 MIM: 610337 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MRPS24 - mitochondrial ribosomal protein S24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
URGCP - upregulator of cell proliferation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077663.2 | 2538 | Silent Mutation | CAC,CAT | H,H 683 | NP_001071131.1 | |
NM_001077664.2 | 2538 | Silent Mutation | CAC,CAT | H,H 640 | NP_001071132.1 | |
NM_001290075.1 | 2538 | Silent Mutation | CAC,CAT | H,H 640 | NP_001277004.1 | |
NM_001290076.1 | 2538 | Silent Mutation | CAC,CAT | H,H 640 | NP_001277005.1 | |
NM_017920.4 | 2538 | Silent Mutation | CAC,CAT | H,H 674 | NP_060390.3 |
URGCP-MRPS24 - URGCP-MRPS24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204871.1 | 2538 | Intron | NP_001191800.1 |