Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613921 MIM: 609360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPJ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPJ - lipase family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010939.2 | Intron | NP_001010939.2 | ||||
XM_005269542.3 | Intron | XP_005269599.1 | ||||
XM_006717635.2 | Intron | XP_006717698.1 | ||||
XM_011539314.2 | Intron | XP_011537616.1 | ||||
XM_011539315.2 | Intron | XP_011537617.1 | ||||
XM_011539317.2 | Intron | XP_011537619.1 | ||||
XM_011539318.2 | Intron | XP_011537620.1 | ||||
XM_011539319.1 | Intron | XP_011537621.1 | ||||
XM_011539320.2 | Intron | XP_011537622.1 | ||||
XM_017015740.1 | Intron | XP_016871229.1 |
RNLS - renalase, FAD dependent amine oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |