Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612790 | ||||||||||||||||||||
Literature Links: |
ARHGEF28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF28 - Rho guanine nucleotide exchange factor 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080479.2 | Intron | NP_001073948.2 | ||||
NM_001177693.1 | Intron | NP_001171164.1 | ||||
NM_001244364.1 | Intron | NP_001231293.1 | ||||
XM_005248568.4 | Intron | XP_005248625.1 | ||||
XM_011543546.1 | Intron | XP_011541848.1 | ||||
XM_011543547.2 | Intron | XP_011541849.1 | ||||
XM_017009702.1 | Intron | XP_016865191.1 | ||||
XM_017009703.1 | Intron | XP_016865192.1 |