Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603219 | ||||||||||||||||||||||||||||||||
Literature Links: |
KCNK2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
KCNK2 - potassium two pore domain channel subfamily K member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017424.2 | 249 | Intron | NP_001017424.1 | |||
NM_001017425.2 | 249 | Intron | NP_001017425.2 | |||
NM_014217.3 | 249 | Intron | NP_055032.1 | |||
XM_011509522.2 | 249 | Intron | XP_011507824.1 | |||
XM_011509523.1 | 249 | Intron | XP_011507825.1 | |||
XM_011509524.2 | 249 | Intron | XP_011507826.1 | |||
XM_017001248.1 | 249 | UTR 5 | XP_016856737.1 | |||
XM_017001249.1 | 249 | UTR 5 | XP_016856738.1 |
Set Membership: |
HapMap |