Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603219 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNK2 - potassium two pore domain channel subfamily K member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017424.2 | Intron | NP_001017424.1 | ||||
NM_001017425.2 | Intron | NP_001017425.2 | ||||
NM_014217.3 | Intron | NP_055032.1 | ||||
XM_011509522.2 | Intron | XP_011507824.1 | ||||
XM_011509523.1 | Intron | XP_011507825.1 | ||||
XM_011509524.2 | Intron | XP_011507826.1 | ||||
XM_017001248.1 | Intron | XP_016856737.1 | ||||
XM_017001249.1 | Intron | XP_016856738.1 |
Set Membership: |
HapMap |