Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC4A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC4A4 - solute carrier family 4 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098484.2 | Intron | NP_001091954.1 | ||||
NM_001134742.1 | Intron | NP_001128214.1 | ||||
NM_003759.3 | Intron | NP_003750.1 | ||||
XM_011532390.2 | Intron | XP_011530692.1 | ||||
XM_017008792.1 | Intron | XP_016864281.1 | ||||
XM_017008793.1 | Intron | XP_016864282.1 |