Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AMER3 - APC membrane recruitment protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105193.1 | 803 | Intron | NP_001098663.1 | |||
NM_001105194.1 | 803 | Intron | NP_001098664.1 | |||
NM_001105195.1 | 803 | Intron | NP_001098665.1 | |||
NM_152698.2 | 803 | Intron | NP_689911.2 | |||
XM_011510793.2 | 803 | UTR 5 | XP_011509095.1 |
POTEI - POTE ankyrin domain family member I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277406.1 | 803 | Intron | NP_001264335.1 | |||
XM_011511670.2 | 803 | Intron | XP_011509972.1 | |||
XM_017004732.1 | 803 | Intron | XP_016860221.1 | |||
XM_017004733.1 | 803 | Intron | XP_016860222.1 | |||
XM_017004734.1 | 803 | Intron | XP_016860223.1 | |||
XM_017004735.1 | 803 | Intron | XP_016860224.1 |
Set Membership: |
HapMap Validated |