Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCGGTCACTTCTCGTCTCAGC[G/T]CTTTCTTTGCTTCTTGGTTTGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602684 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADGRB3 - adhesion G protein-coupled receptor B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001704.2 | 5 | Intron | NP_001695.1 | |||
XM_005248752.2 | 5 | UTR 5 | XP_005248809.1 | |||
XM_006715534.3 | 5 | UTR 5 | XP_006715597.1 | |||
XM_011536009.2 | 5 | Intron | XP_011534311.1 | |||
XM_011536010.2 | 5 | UTR 5 | XP_011534312.1 | |||
XM_011536011.2 | 5 | UTR 5 | XP_011534313.1 | |||
XM_011536012.2 | 5 | UTR 5 | XP_011534314.1 | |||
XM_017011140.1 | 5 | UTR 5 | XP_016866629.1 | |||
XM_017011141.1 | 5 | Intron | XP_016866630.1 | |||
XM_017011142.1 | 5 | UTR 5 | XP_016866631.1 | |||
XM_017011143.1 | 5 | UTR 5 | XP_016866632.1 | |||
XM_017011144.1 | 5 | UTR 5 | XP_016866633.1 |
LOC101928307 - uncharacterized LOC101928307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |