Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF420 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF420 - zinc finger protein 420 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006723038.3 | Intron | XP_006723101.1 | ||||
XM_006723039.3 | Intron | XP_006723102.1 | ||||
XM_011526503.2 | Intron | XP_011524805.1 | ||||
XM_011526504.2 | Intron | XP_011524806.1 | ||||
XM_011526505.2 | Intron | XP_011524807.1 | ||||
XM_011526507.2 | Intron | XP_011524809.1 | ||||
XM_011526508.2 | Intron | XP_011524810.1 | ||||
XM_011526510.2 | Intron | XP_011524812.1 | ||||
XM_011526511.2 | Intron | XP_011524813.1 | ||||
XM_017026336.1 | Intron | XP_016881825.1 | ||||
XM_017026337.1 | Intron | XP_016881826.1 | ||||
XM_017026338.1 | Intron | XP_016881827.1 | ||||
XM_017026339.1 | Intron | XP_016881828.1 | ||||
XM_017026340.1 | Intron | XP_016881829.1 |
Set Membership: |
HapMap |