Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612779 | ||||||||||||||||||||
Literature Links: |
DPYD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPYD - dihydropyrimidine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000110.3 | 3891 | UTR 3 | NP_000101.2 | |||
NM_001160301.1 | 3891 | Intron | NP_001153773.1 | |||
XM_005270562.3 | 3891 | UTR 3 | XP_005270619.2 | |||
XM_006710397.3 | 3891 | Intron | XP_006710460.1 | |||
XM_017000507.1 | 3891 | UTR 3 | XP_016855996.1 | |||
XM_017000508.1 | 3891 | UTR 3 | XP_016855997.1 | |||
XM_017000509.1 | 3891 | UTR 3 | XP_016855998.1 | |||
XM_017000510.1 | 3891 | UTR 3 | XP_016855999.1 |