Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAGTATTGCCTATCTCTTTCAA[C/G]AGAGTGACATATTATGCCCGATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARMIL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARMIL1 - capping protein regulator and myosin 1 linker 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173977.1 | Intron | NP_001167448.1 | ||||
NM_017640.5 | Intron | NP_060110.4 | ||||
XM_005249218.1 | Intron | XP_005249275.1 | ||||
XM_017011008.1 | Intron | XP_016866497.1 | ||||
XM_017011009.1 | Intron | XP_016866498.1 | ||||
XM_017011010.1 | Intron | XP_016866499.1 | ||||
XM_017011011.1 | Intron | XP_016866500.1 | ||||
XM_017011012.1 | Intron | XP_016866501.1 | ||||
XM_017011013.1 | Intron | XP_016866502.1 | ||||
XM_017011014.1 | Intron | XP_016866503.1 | ||||
XM_017011015.1 | Intron | XP_016866504.1 | ||||
XM_017011016.1 | Intron | XP_016866505.1 | ||||
XM_017011017.1 | Intron | XP_016866506.1 |
LOC105375106 - uncharacterized LOC105375106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |