Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603897 MIM: 616104 | ||||||||||||||||||||
Literature Links: |
MATN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MATN4 - matrilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003833.4 | Intron | NP_003824.2 | ||||
NM_030590.3 | Intron | NP_085080.1 | ||||
NM_030592.3 | Intron | NP_085095.1 | ||||
XM_005260597.1 | Intron | XP_005260654.1 | ||||
XM_017028113.1 | Intron | XP_016883602.1 | ||||
XM_017028114.1 | Intron | XP_016883603.1 | ||||
XM_017028115.1 | Intron | XP_016883604.1 |
RBPJL - recombination signal binding protein for immunoglobulin kappa J region like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281448.1 | Intron | NP_001268377.1 | ||||
NM_001281449.1 | Intron | NP_001268378.1 | ||||
NM_014276.3 | Intron | NP_055091.2 | ||||
XM_011528521.2 | Intron | XP_011526823.1 | ||||
XM_011528522.2 | Intron | XP_011526824.1 | ||||
XM_011528523.2 | Intron | XP_011526825.1 |