Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602842 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMNN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMNN - geminin, DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251989.1 | 753 | Intron | NP_001238918.1 | |||
NM_001251990.1 | 753 | Intron | NP_001238919.1 | |||
NM_001251991.1 | 753 | Intron | NP_001238920.1 | |||
NM_015895.4 | 753 | Intron | NP_056979.1 | |||
XM_005249159.1 | 753 | Intron | XP_005249216.1 | |||
XM_011514651.2 | 753 | UTR 5 | XP_011512953.1 |