Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 609642 | ||||||||||||||||||||||||||
Literature Links: |
TARP PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TARP - TCR gamma alternate reading frame protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003799.1 | Intron | NP_001003799.1 | ||||
NM_001003806.1 | Intron | NP_001003806.1 | ||||
XM_011515407.2 | Intron | XP_011513709.1 | ||||
XM_017012226.1 | Intron | XP_016867715.1 | ||||
XM_017012227.1 | Intron | XP_016867716.1 | ||||
XM_017012228.1 | Intron | XP_016867717.1 | ||||
XM_017012229.1 | Intron | XP_016867718.1 | ||||
XM_017012230.1 | Intron | XP_016867719.1 |