Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609240 MIM: 604682 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GSG2 - germ cell associated 2, haspin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGAE - integrin subunit alpha E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002208.4 | Intron | NP_002199.3 | ||||
XM_011523823.2 | Intron | XP_011522125.1 | ||||
XM_011523825.2 | Intron | XP_011522127.1 | ||||
XM_011523827.2 | Intron | XP_011522129.1 | ||||
XM_011523828.2 | Intron | XP_011522130.1 | ||||
XM_017024586.1 | Intron | XP_016880075.1 | ||||
XM_017024587.1 | Intron | XP_016880076.1 |
Set Membership: |
HapMap |