Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611470 MIM: 601585 | ||||||||||||||||||||
Literature Links: |
LGSN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGSN - lengsin, lens protein with glutamine synthetase domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTP4A1 - protein tyrosine phosphatase type IVA, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003463.4 | Intron | NP_003454.1 | ||||
XM_011536111.1 | Intron | XP_011534413.1 | ||||
XM_011536112.1 | Intron | XP_011534414.1 | ||||
XM_017011270.1 | Intron | XP_016866759.1 | ||||
XM_017011271.1 | Intron | XP_016866760.1 | ||||
XM_017011272.1 | Intron | XP_016866761.1 |