Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGGAAGGCAGGCGTGGTCAGC[A/G]AGGACTGGCTTCTGCGTGAGCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171840 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PFKP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PFKP - phosphofructokinase, platelet | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITRM1 - pitrilysin metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242307.1 | Intron | NP_001229236.1 | ||||
NM_001242309.1 | Intron | NP_001229238.1 | ||||
NM_014889.3 | Intron | NP_055704.2 | ||||
XM_005252345.3 | Intron | XP_005252402.1 | ||||
XM_017015470.1 | Intron | XP_016870959.1 | ||||
XM_017015471.1 | Intron | XP_016870960.1 | ||||
XM_017015472.1 | Intron | XP_016870961.1 | ||||
XM_017015473.1 | Intron | XP_016870962.1 |
PITRM1-AS1 - PITRM1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD142 - small nucleolar RNA, C/D box 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |