Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGTTTGGAATGGAGCAGTGCAGG[A/T]GGGATGACTATGACCCTGACGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125265 MIM: 182175 MIM: 601079 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
REEP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
REEP5 - receptor accessory protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005669.4 | 1188 | Intron | NP_005660.4 | |||
XM_017009844.1 | 1188 | Intron | XP_016865333.1 |
SRP19 - signal recognition particle 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204193.1 | 1188 | Intron | NP_001191122.1 | |||
NM_001204194.1 | 1188 | Intron | NP_001191123.1 | |||
NM_001204196.1 | 1188 | Intron | NP_001191125.1 | |||
NM_001204199.1 | 1188 | UTR 3 | NP_001191128.1 | |||
NM_003135.2 | 1188 | Intron | NP_003126.1 |
ZRSR1 - zinc finger CCCH-type, RNA binding motif and serine/arginine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |