Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607321 MIM: 114250 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP1A4 - ATPase Na+/K+ transporting subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CASQ1 - calsequestrin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001231.4 | Intron | NP_001222.3 |
LOC729867 - uncharacterized LOC729867 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |