Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606264 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC7A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLEC7A - C-type lectin domain family 7 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022570.4 | 696 | UTR 3 | NP_072092.2 | |||
NM_197947.2 | 696 | UTR 3 | NP_922938.1 | |||
NM_197948.2 | 696 | UTR 3 | NP_922939.1 | |||
NM_197949.2 | 696 | UTR 3 | NP_922940.1 | |||
NM_197950.2 | 696 | UTR 3 | NP_922941.1 | |||
NM_197954.2 | 696 | Intron | NP_922945.1 | |||
XM_006719135.3 | 696 | Intron | XP_006719198.1 | |||
XM_017019822.1 | 696 | UTR 3 | XP_016875311.1 | |||
XM_017019823.1 | 696 | UTR 3 | XP_016875312.1 |
Set Membership: |
HapMap Validated |