Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCACCTCTACCTTCTTCATAAA[C/T]TTTTAAGATGAATTGGTAATTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUT - deoxyuridine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025248.1 | Intron | NP_001020419.1 | ||||
NM_001025249.1 | Intron | NP_001020420.1 | ||||
NM_001948.3 | Intron | NP_001939.1 | ||||
XM_011521315.2 | Intron | XP_011519617.1 | ||||
XM_017021988.1 | Intron | XP_016877477.1 |