Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104613 MIM: 172480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT6A - chaperonin containing TCP1 subunit 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009186.1 | Intron | NP_001009186.1 | ||||
NM_001762.3 | Intron | NP_001753.1 |
PSPH - phosphoserine phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004577.3 | Intron | NP_004568.2 | ||||
XM_005271773.1 | Intron | XP_005271830.1 | ||||
XM_005271774.1 | Intron | XP_005271831.1 | ||||
XM_005271775.1 | Intron | XP_005271832.1 | ||||
XM_005271776.1 | Intron | XP_005271833.1 | ||||
XM_006715760.1 | Intron | XP_006715823.1 | ||||
XM_006715761.1 | Intron | XP_006715824.1 | ||||
XM_011515461.1 | Intron | XP_011513763.1 | ||||
XM_017012466.1 | Intron | XP_016867955.1 | ||||
XM_017012467.1 | Intron | XP_016867956.1 | ||||
XM_017012468.1 | Intron | XP_016867957.1 | ||||
XM_017012469.1 | Intron | XP_016867958.1 |
SNORA15 - small nucleolar RNA, H/ACA box 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |