Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCAGGCTGCTTCGTAGCGGGC[A/G]TCGGCTGCGGACTTCCCTTCCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615099 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERFE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERFE - erythroferrone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ILKAP - ILK associated serine/threonine phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030768.2 | 1070 | Silent Mutation | NP_110395.1 | |||
XM_005246106.1 | 1070 | Silent Mutation | XP_005246163.1 | |||
XM_006712784.1 | 1070 | Silent Mutation | XP_006712847.1 | |||
XM_011511946.2 | 1070 | Silent Mutation | XP_011510248.1 | |||
XM_017005056.1 | 1070 | Missense Mutation | XP_016860545.1 | |||
XM_017005057.1 | 1070 | Silent Mutation | XP_016860546.1 | |||
XM_017005058.1 | 1070 | Silent Mutation | XP_016860547.1 |