Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120250 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL6A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL6A3 - collagen type VI alpha 3 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004369.3 | 8005 | UTR 3 | NP_004360.2 | |||
NM_057164.4 | 8005 | Intron | NP_476505.3 | |||
NM_057165.4 | 8005 | Intron | NP_476506.3 | |||
NM_057166.4 | 8005 | UTR 3 | NP_476507.3 | |||
NM_057167.3 | 8005 | UTR 3 | NP_476508.2 | |||
XM_005246065.1 | 8005 | UTR 3 | XP_005246122.1 | |||
XM_005246066.1 | 8005 | UTR 3 | XP_005246123.1 | |||
XM_006712253.1 | 8005 | UTR 3 | XP_006712316.1 | |||
XM_011510574.1 | 8005 | UTR 3 | XP_011508876.1 | |||
XM_017003303.1 | 8005 | UTR 3 | XP_016858792.1 | |||
XM_017003304.1 | 8005 | UTR 3 | XP_016858793.1 |
Set Membership: |
HapMap |