Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAATCATCATAGCTACATGTC[G/T]TGGCTGTTGTGCCTGGCCAGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRS2 - MRS2, magnesium transporter | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286264.1 | Intron | NP_001273193.1 | ||||
NM_001286265.1 | Intron | NP_001273194.1 | ||||
NM_001286266.1 | Intron | NP_001273195.1 | ||||
NM_020662.3 | Intron | NP_065713.1 | ||||
XM_005249242.1 | Intron | XP_005249299.1 |