Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTCAGACTCAGGTTCTTGAAT[C/G]TGGTCTCCCATGGGCCGACTGTTCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604358 | |||||||||||||||||||||||
Literature Links: |
STAG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
STAG1 - stromal antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005862.2 | 2260 | Missense Mutation | CAC,CAG | H,Q 1132 | NP_005853.2 | |
XM_011512329.2 | 2260 | Missense Mutation | CAC,CAG | H,Q 995 | XP_011510631.1 | |
XM_011512331.2 | 2260 | Missense Mutation | CAC,CAG | H,Q 995 | XP_011510633.1 | |
XM_017005523.1 | 2260 | Missense Mutation | CAC,CAG | H,Q 995 | XP_016861012.1 | |
XM_017005524.1 | 2260 | Missense Mutation | CAC,CAG | H,Q 906 | XP_016861013.1 | |
XM_017005525.1 | 2260 | Missense Mutation | CAC,CAG | H,Q 695 | XP_016861014.1 |