Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCTCAACTTTAGAGGCCCTAA[A/T]CCCCAAGCAGTTGGGCCCAAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF84 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF84 - zinc finger protein 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127372.2 | Intron | NP_001120844.1 | ||||
NM_001289971.1 | Intron | NP_001276900.1 | ||||
NM_001289972.1 | Intron | NP_001276901.1 | ||||
NM_003428.5 | Intron | NP_003419.3 | ||||
XM_005266185.1 | Intron | XP_005266242.1 | ||||
XM_005266186.1 | Intron | XP_005266243.1 | ||||
XM_011534832.1 | Intron | XP_011533134.1 |