Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTTTCTGTCCTGTGCAAATCC[A/T]CTGTTATCTCCTGAGGCTGCACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605422 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF350 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF350 - zinc finger protein 350 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021632.3 | 1890 | Missense Mutation | AGA,AGT | R,S 501 | NP_067645.3 | |
XM_011527196.1 | 1890 | Missense Mutation | AGA,AGT | R,S 465 | XP_011525498.1 | |
XM_017027094.1 | 1890 | Missense Mutation | AGA,AGT | R,S 543 | XP_016882583.1 | |
XM_017027095.1 | 1890 | Missense Mutation | AGA,AGT | R,S 543 | XP_016882584.1 | |
XM_017027096.1 | 1890 | Missense Mutation | AGA,AGT | R,S 543 | XP_016882585.1 | |
XM_017027097.1 | 1890 | Missense Mutation | AGA,AGT | R,S 507 | XP_016882586.1 | |
XM_017027098.1 | 1890 | Missense Mutation | AGA,AGT | R,S 501 | XP_016882587.1 | |
XM_017027099.1 | 1890 | Missense Mutation | AGA,AGT | R,S 501 | XP_016882588.1 | |
XM_017027100.1 | 1890 | Missense Mutation | AGA,AGT | R,S 501 | XP_016882589.1 | |
XM_017027101.1 | 1890 | Missense Mutation | AGA,AGT | R,S 275 | XP_016882590.1 | |
XM_017027102.1 | 1890 | Missense Mutation | AGA,AGT | R,S 275 | XP_016882591.1 | |
XM_017027103.1 | 1890 | Missense Mutation | AGA,AGT | R,S 275 | XP_016882592.1 | |
XM_017027104.1 | 1890 | Missense Mutation | AGA,AGT | R,S 275 | XP_016882593.1 |
ZNF350-AS1 - ZNF350 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |