Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCACACCATCATCAGTTTCCGA[C/T]GTTCTCGGGGAGTTGCTGGAGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFIL3 - nuclear factor, interleukin 3 regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289999.1 | 1355 | Silent Mutation | ACA,ACG | T,T 274 | NP_001276928.1 | |
NM_001290000.1 | 1355 | Silent Mutation | ACA,ACG | T,T 274 | NP_001276929.1 | |
NM_005384.2 | 1355 | Silent Mutation | ACA,ACG | T,T 274 | NP_005375.2 | |
XM_017014743.1 | 1355 | Silent Mutation | ACA,ACG | T,T 274 | XP_016870232.1 | |
XM_017014744.1 | 1355 | Silent Mutation | ACA,ACG | T,T 274 | XP_016870233.1 |