Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612810 MIM: 608786 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRFN4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRFN4 - leucine rich repeat and fibronectin type III domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024036.4 | 1239 | Missense Mutation | GCG,GTG | A,V 340 | NP_076941.2 | |
XM_005274239.3 | 1239 | Missense Mutation | GCG,GTG | A,V 340 | XP_005274296.1 |
PC - pyruvate carboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000920.3 | 1239 | Intron | NP_000911.2 | |||
NM_001040716.1 | 1239 | Intron | NP_001035806.1 | |||
NM_022172.2 | 1239 | Intron | NP_071504.2 | |||
XM_005274031.4 | 1239 | Intron | XP_005274088.1 | |||
XM_005274032.4 | 1239 | Intron | XP_005274089.1 | |||
XM_006718578.3 | 1239 | Intron | XP_006718641.1 | |||
XM_006718579.3 | 1239 | Intron | XP_006718642.1 | |||
XM_011545086.2 | 1239 | Intron | XP_011543388.1 | |||
XM_011545087.2 | 1239 | Intron | XP_011543389.1 | |||
XM_017017868.1 | 1239 | Intron | XP_016873357.1 | |||
XM_017017869.1 | 1239 | Intron | XP_016873358.1 | |||
XM_017017870.1 | 1239 | Intron | XP_016873359.1 | |||
XM_017017871.1 | 1239 | Intron | XP_016873360.1 | |||
XM_017017872.1 | 1239 | Intron | XP_016873361.1 |
Set Membership: |
HapMap Validated |