Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCAGGCTTGTAGATCTGAGCC[A/G]CTGAGATCTAGGACATGTGCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100850 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACO2 - aconitase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098.2 | 2559 | Intron | NP_001089.1 | |||
XM_017028812.1 | 2559 | Intron | XP_016884301.1 |
POLR3H - RNA polymerase III subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018050.3 | 2559 | UTR 3 | NP_001018060.1 | |||
NM_001018052.3 | 2559 | UTR 3 | NP_001018062.1 | |||
NM_001282884.1 | 2559 | UTR 3 | NP_001269813.1 | |||
NM_001282885.1 | 2559 | UTR 3 | NP_001269814.1 | |||
NM_138338.4 | 2559 | UTR 3 | NP_612211.1 | |||
XM_011529993.1 | 2559 | Intron | XP_011528295.1 | |||
XM_017028641.1 | 2559 | Intron | XP_016884130.1 |
Set Membership: |
HapMap |